Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546873.2 | XP_005546930.1 | 7 - 8 | 2122 | 115 | |
| XM_005546875.2 | XP_005546932.1 | 7 - 8 | 2588 | 115 | ||
More Information
Set Membership: |
|

CDCP1
Genomic Map