Assay Details
| Gene Symbol: C8H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564438.2 | XP_005564495.1 | 3 - 4 | 512 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H8orf33| Gene Symbol: C8H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564438.2 | XP_005564495.1 | 3 - 4 | 512 | 105 | |
Set Membership: |
|