Assay Details
| Gene Symbol: HIVEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543760.2 | XP_005543817.1 | 2 - 3 | 7840 | 121 | |
| XM_005543762.2 | XP_005543819.1 | 5 - 6 | 6554 | 121 | ||
| XM_005543763.2 | XP_005543820.1 | 2 - 3 | 7173 | 121 | ||
| XM_015437593.1 | XP_015293079.1 | 4 - 5 | 7361 | 121 | ||
| XM_015437609.1 | XP_015293095.1 | 5 - 6 | 6021 | 121 | ||
| XM_015437619.1 | XP_015293105.1 | 4 - 5 | 7361 | 121 | ||
| GenBank mRNA | AB179018.1 | - | 1 - 2 | 1213 | 121 | |
More Information
Set Membership: |
|

HIVEP3
Genomic Map