Assay Details
| Gene Symbol: IQCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574709.2 | XP_005574766.1 | 4 - 5 | 920 | 131 | |
| XM_015433008.1 | XP_015288494.1 | 4 - 5 | 791 | 131 | ||
| XM_015433009.1 | XP_015288495.1 | 4 - 5 | 791 | 131 | ||
| XM_015433010.1 | XP_015288496.1 | 4 - 5 | 793 | 131 | ||
| XM_015433012.1 | XP_015288498.1 | 4 - 5 | 805 | 131 | ||
More Information
Set Membership: |
|

IQCA1
Genomic Map