Assay Details
| Gene Symbol: PDZD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579914.2 | XP_005579971.1 | 6 - 7 | 653 | 125 | |
| XM_005579915.2 | XP_005579972.1 | 6 - 7 | 675 | 125 | ||
More Information
Set Membership: |
|

PDZD3
Genomic Map