Assay Details
| Gene Symbol: SETD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592117.2 | XP_005592174.1 | 5 - 6 | 865 | 106 | |
| XM_005592118.2 | XP_005592175.1 | 6 - 7 | 788 | 106 | ||
| XM_015442813.1 | XP_015298299.1 | 4 - 5 | 1851 | 106 | ||
| XM_015442814.1 | XP_015298300.1 | 5 - 6 | 1775 | 106 | ||
| XM_015442815.1 | XP_015298301.1 | 5 - 6 | 1400 | 106 | ||
More Information
Set Membership: |
|

SETD6
Genomic Map