Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561106.2 | XP_005561163.1 | 8 - 9 | 1855 | 96 | |
| XM_005561107.2 | XP_005561164.1 | 8 - 9 | 1857 | 96 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map