Assay Details
| Gene Symbol: UCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580603.2 | XP_005580660.1 | 1 - 2 | 210 | 130 | |
| XM_005580605.2 | XP_005580662.1 | 1 - 2 | 207 | 130 | ||
| GenBank mRNA | AB169892.1 | - | 1 - 2 | 137 | 130 | |
| AB170445.1 | - | 1 - 2 | 159 | 130 | ||
More Information
Set Membership: |
|

UCK1
Genomic Map