Assay Details
| Gene Symbol: GORASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546691.2 | XP_005546748.1 | 5 - 6 | 857 | 93 | |
| XM_005546692.2 | XP_005546749.1 | 5 - 6 | 857 | 93 | ||
| XM_005546693.2 | XP_005546750.1 | 5 - 6 | 857 | 93 | ||
| XM_005546694.2 | XP_005546751.1 | 5 - 6 | 720 | 93 | ||
| XM_005546695.2 | XP_005546752.1 | 5 - 6 | 858 | 93 | ||
| XM_005546697.2 | XP_005546754.1 | 3 - 4 | 427 | 93 | ||
More Information
Set Membership: |
|

GORASP1
Genomic Map