Assay Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319445.1 | NP_001306374.1 | 2 - 3 | 1052 | 119 | |
| GenBank mRNA | AB074449.1 | - | 2 - 3 | 1052 | 119 | |
More Information
Set Membership: |
|

HIPK4
Genomic Map