Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542010.2 | XP_005542067.1 | 4 - 5 | 1268 | 149 | |
| XM_005542012.2 | XP_005542069.1 | 5 - 6 | 768 | 149 | ||
| XM_015457779.1 | XP_015313265.1 | 6 - 7 | 878 | 149 | ||
| XM_015457784.1 | XP_015313270.1 | 6 - 7 | 572 | 149 | ||
| XM_015457786.1 | XP_015313272.1 | 5 - 6 | 469 | 149 | ||
More Information
Set Membership: |
|

CIART
Genomic Map