Assay Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579211.2 | XP_005579268.1 | 2 - 3 | 236 | 93 | |
| XM_015435550.1 | XP_015291036.1 | 3 - 4 | 323 | 93 | ||
More Information
Set Membership: |
|

DDIAS
Genomic Map