Assay Details
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577152.1 | XP_005577209.1 | 6 - 7 | 806 | 106 | |
XM_005577154.1 | XP_005577211.1 | 5 - 6 | 734 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
