Assay Details
| Gene Symbol: LOC102137822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565979.2 | XP_005566036.1 | 1 - 2 | 184 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005565979.2 transcripts.
Set Membership: |
|

LOC102137822
Genomic Map