Assay Details
| Gene Symbol: LOC102137822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565976.2 | XP_005566033.1 | 4 - 5 | 333 | 140 | |
| XM_005565977.2 | XP_005566034.1 | 4 - 5 | 574 | 140 | ||
| XM_005565978.2 | XP_005566035.1 | 4 - 5 | 240 | 140 | ||
| XM_005565979.2 | XP_005566036.1 | 4 - 5 | 400 | 140 | ||
| XM_005565980.2 | XP_005566037.1 | 4 - 5 | 335 | 140 | ||
More Information
Set Membership: |
|

LOC102137822
Genomic Map