Assay Details
| Gene Symbol: C14H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579533.2 | XP_005579590.1 | 8 - 9 | 808 | 87 | |
| XM_005579535.1 | XP_005579592.1 | 7 - 8 | 800 | 87 | ||
| XM_005579537.2 | XP_005579594.1 | 6 - 7 | 713 | 87 | ||
| XM_005579538.2 | XP_005579595.1 | 6 - 7 | 672 | 87 | ||
| XM_005579539.1 | XP_005579596.1 | 5 - 6 | 644 | 87 | ||
| XM_005579541.2 | XP_005579598.1 | 5 - 6 | 653 | 87 | ||
More Information
Set Membership: |
|

C14H11orf65
Genomic Map