Assay Details
| Gene Symbol: PARP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547983.2 | XP_005548040.2 | 8 - 9 | 2978 | 118 | |
| XM_015446353.1 | XP_015301839.1 | 7 - 8 | 2828 | 118 | ||
More Information
Set Membership: |
|

PARP15
Genomic Map