Assay Details
| Gene Symbol: GLIS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581803.2 | XP_005581860.1 | 10 - 11 | 2866 | 83 | |
| XM_005581804.2 | XP_005581861.1 | 10 - 11 | 4584 | 83 | ||
| XM_005581807.2 | XP_005581864.1 | 9 - 10 | 3508 | 83 | ||
| XM_005581810.2 | XP_005581867.1 | 9 - 10 | 2343 | 83 | ||
| XM_005581812.2 | XP_005581869.1 | 8 - 9 | 2228 | 83 | ||
| XM_015437223.1 | XP_015292709.1 | 9 - 10 | 10514 | 83 | ||
| XM_015437225.1 | XP_015292711.1 | 9 - 10 | 2658 | 83 | ||
More Information
Set Membership: |
|

GLIS3
Genomic Map