Assay Details
| Gene Symbol: LOC102136101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434334.1 | XP_015289820.1 | 4 - 5 | 463 | 135 | |
| XM_015434335.1 | XP_015289821.1 | 4 - 5 | 463 | 135 | ||
More Information
Set Membership: |
|

LOC102136101
Genomic Map