Assay Details
| Gene Symbol: LYPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589425.2 | XP_005589482.1 | 3 - 4 | 1382 | 141 | |
| XM_015442089.1 | XP_015297575.1 | 3 - 4 | 1451 | 141 | ||
More Information
Set Membership: |
|

LYPD4
Genomic Map