Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596053.2 | XP_005596110.1 | 8 - 9 | 1064 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEXD| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596053.2 | XP_005596110.1 | 8 - 9 | 1064 | 64 | |
Set Membership: |
|