Assay Details
| Gene Symbol: LOC102132535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555097.2 | XP_005555154.1 | 1 - 2 | 501 | 127 | |
| XM_005555098.2 | XP_005555155.1 | 1 - 2 | 502 | 127 | ||
| XM_005555099.2 | XP_005555156.1 | 1 - 2 | 504 | 127 | ||
More Information
Set Membership: |
|

LOC102132535
Genomic Map