Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433786.1 | XP_015289272.1 | 5 - 6 | 568 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPATCH11| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433786.1 | XP_015289272.1 | 5 - 6 | 568 | 146 | |
Set Membership: |
|