Assay Details
| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578813.2 | XP_005578870.1 | 3 - 4 | 1184 | 135 | |
| XM_015435230.1 | XP_015290716.1 | 3 - 4 | 1359 | 135 | ||
More Information
Set Membership: |
|

CCKBR
Genomic Map