Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549361.2 | XP_005549418.1 | 5 - 6 | 1118 | 81 | |
XM_015447210.1 | XP_015302696.1 | 4 - 5 | 1259 | 81 | ||
XM_015447211.1 | XP_015302697.1 | 3 - 4 | 1907 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
