Assay Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584058.2 | XP_005584115.1 | 8 - 9 | 1314 | 71 | |
| XM_005584060.2 | XP_005584117.1 | 7 - 8 | 1287 | 71 | ||
More Information
Set Membership: |
|

GSDMA
Genomic Map