Assay Details
| Gene Symbol: GPR141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549773.2 | XP_005549830.1 | 5 | 1172 | 132 | |
| XM_005549774.2 | XP_005549831.1 | 4 | 1077 | 132 | ||
| XM_005549775.2 | XP_005549832.1 | 4 | 1073 | 132 | ||
| XM_005549776.2 | XP_005549833.1 | 3 | 951 | 132 | ||
| XM_005549777.2 | XP_005549834.1 | 2 | 1017 | 132 | ||
| XM_005549778.2 | XP_005549835.1 | 2 | 857 | 132 | ||
| XM_015447518.1 | XP_015303004.1 | 4 | 2971 | 132 | ||
| XM_015447519.1 | XP_015303005.1 | 6 | 1322 | 132 | ||
| XM_015447520.1 | XP_015303006.1 | 5 | 5782 | 132 | ||
| XM_015447521.1 | XP_015303007.1 | 3 | 978 | 132 | ||
More Information
Set Membership: |
|

GPR141
Genomic Map