Assay Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443789.1 | XP_015299275.1 | 2 - 3 | 251 | 75 | |
| XM_015443790.1 | XP_015299276.1 | 2 - 3 | 251 | 75 | ||
More Information
Set Membership: |
|

FAAP20
Genomic Map