Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582581.2 | XP_005582638.1 | 1 - 2 | 364 | 159 | |
| GenBank mRNA | AB171167.1 | - | 1 - 2 | 198 | 160 | |
More Information
Set Membership: |
|

VMO1
Genomic Map