Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549535.2 | XP_005549592.1 | 6 - 7 | 1251 | 140 | |
| XM_005549537.2 | XP_005549594.1 | 6 - 7 | 1233 | 140 | ||
| XM_015447384.1 | XP_015302870.1 | 7 - 8 | 1489 | 140 | ||
| XM_015447385.1 | XP_015302871.1 | 7 - 8 | 1157 | 140 | ||
| XM_015447386.1 | XP_015302872.1 | 3 - 4 | 848 | 140 | ||
More Information
Set Membership: |
|

ZNF713
Genomic Map