Assay Details
| Gene Symbol: RIMS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569097.2 | XP_005569154.1 | 4 - 5 | 582 | 58 | |
| XM_005569099.2 | XP_005569156.1 | 4 - 5 | 582 | 58 | ||
More Information
Set Membership: |
|

RIMS4
Genomic Map