Assay Details
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565961.2 | XP_005566018.1 | 2 - 3 | 738 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
