Assay Details
| Gene Symbol: PRSS55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562607.2 | XP_005562664.1 | 1 - 2 | 188 | 146 | |
| XM_005562611.2 | XP_005562668.1 | 1 - 2 | 472 | 146 | ||
More Information
Set Membership: |
|

PRSS55
Genomic Map