Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588022.1 | XP_005588079.1 | 2 - 3 | 337 | 151 | |
| XM_005588023.2 | XP_005588080.1 | 2 - 3 | 377 | 151 | ||
| XM_005588025.2 | XP_005588082.1 | 2 - 3 | 408 | 151 | ||
| XM_005588026.2 | XP_005588083.1 | 2 - 3 | 277 | 151 | ||
| XM_005588027.2 | XP_005588084.1 | 2 - 3 | 254 | 151 | ||
| XM_005588028.2 | XP_005588085.1 | 2 - 3 | 127 | 151 | ||
| XM_015440543.1 | XP_015296029.1 | 2 - 3 | 250 | 151 | ||
| GenBank mRNA | AB170107.1 | - | 178 | 151 | ||
More Information
Set Membership: |
|

TMEM205
Genomic Map