Assay Details
| Gene Symbol: MN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567714.2 | XP_005567771.1 | 1 | 897 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005567714.2 transcripts.
Set Membership: |
|

MN1
Genomic Map