Assay Details
| Gene Symbol: MIER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543110.1 | XP_005543167.1 | 4 - 5 | 396 | 103 | |
| XM_005543111.1 | XP_005543168.1 | 4 - 5 | 399 | 103 | ||
| XM_005543114.1 | XP_005543171.1 | 4 - 5 | 357 | 103 | ||
| XM_015434467.1 | XP_015289953.1 | 4 - 5 | 265 | 103 | ||
| XM_015434471.1 | XP_015289957.1 | 4 - 5 | 327 | 103 | ||
| XM_015434483.1 | XP_015289969.1 | 4 - 5 | 349 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005543114.1 transcripts.
Set Membership: |
|

MIER1
Genomic Map