Assay Details
| Gene Symbol: ITPRIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574984.2 | XP_005575041.1 | 1 | 212 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005574984.2 transcripts.
Set Membership: |
|

ITPRIPL1
Genomic Map