Assay Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555072.2 | XP_005555129.1 | 3 - 4 | 320 | 79 | |
| XM_005555073.2 | XP_005555130.1 | 2 - 3 | 183 | 79 | ||
| XM_015450395.1 | XP_015305881.1 | 3 - 4 | 293 | 79 | ||
More Information
Set Membership: |
|

EPGN
Genomic Map