Assay Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552780.2 | XP_005552837.1 | 3 | 2468 | 123 | |
| XM_015449330.1 | XP_015304816.1 | 3 | 2642 | 123 | ||
| XM_015449331.1 | XP_015304817.1 | 4 | 2831 | 123 | ||
| XM_015449332.1 | XP_015304818.1 | 4 | 2656 | 123 | ||
More Information
Set Membership: |
|

PTCHD4
Genomic Map