Assay Details
| Gene Symbol: LOC102131535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556828.2 | XP_005556885.1 | 1 | 1475 | 143 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005556828.2 transcripts.
Set Membership: |
|

LOC102131535
Genomic Map