Assay Details
| Gene Symbol: C15H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437040.1 | XP_015292526.1 | 2 | 2856 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H9orf131| Gene Symbol: C15H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437040.1 | XP_015292526.1 | 2 | 2856 | 108 | |
Set Membership: |
|