Assay Details
| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549393.2 | XP_005549450.1 | 1 | 2414 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005549393.2 transcripts.
Set Membership: |
|

CLDN4
Genomic Map