Assay Details
| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015431425.1 | XP_015286911.1 | 1 | 2949 | 120 | |
| XM_015431430.1 | XP_015286916.1 | 1 | 2949 | 120 | ||
More Information
Set Membership: |
|

KCNA3
Genomic Map