Assay Details
| Gene Symbol: CHST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578098.2 | XP_005578155.1 | 1 - 2 | 473 | 93 | |
| XM_005578107.2 | XP_005578164.1 | 1 - 2 | 473 | 93 | ||
| XM_015434864.1 | XP_015290350.1 | 1 - 2 | 473 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005578098.2, XM_005578107.2, XM_015434864.1 transcripts.
Set Membership: |
|

CHST1
Genomic Map