Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565577.2 | XP_005565634.1 | 2 - 3 | 594 | 147 | |
| XM_005565578.2 | XP_005565635.1 | 2 - 3 | 2495 | 147 | ||
More Information
Set Membership: |
|

CHST3
Genomic Map