Assay Details
| Gene Symbol: HSPA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319447.1 | NP_001306376.1 | 1 - 2 | 137 | 135 | |
| GenBank mRNA | AB168570.1 | - | 1 - 2 | 153 | 135 | |
| AB168812.1 | - | 1 - 2 | 140 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001319447.1 transcripts.
Set Membership: |
|

HSPA1L
Genomic Map