Assay Details
| Gene Symbol: KCNA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542407.1 | XP_005542464.1 | 1 | 376 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005542407.1 transcripts.
Set Membership: |
|

KCNA10
Genomic Map