Assay Details
Gene Symbol: ZNF234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589504.2 | XP_005589561.1 | 5 - 6 | 489 | 138 | |
XM_005589505.2 | XP_005589562.1 | 5 - 6 | 494 | 138 | ||
XM_015441130.1 | XP_015296616.1 | 2 - 3 | 677 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441130.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
