Assay Details
| Gene Symbol: SUGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588537.2 | XP_005588594.1 | 2 - 3 | 278 | 124 | |
| XM_015440519.1 | XP_015296005.1 | 2 - 3 | 278 | 124 | ||
More Information
Set Membership: |
|

SUGP1
Genomic Map