Assay Details
| Gene Symbol: LOC102118079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592575.2 | XP_005592632.1 | 2 - 3 | 592 | 122 | |
| XM_015442994.1 | XP_015298480.1 | 2 - 3 | 624 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005592575.2, XM_015442994.1 transcripts.
Set Membership: |
|

LOC102118079
Genomic Map